The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003299.2 | NP_003290.1 | 2 - 3 | 341 | 111 |
GenBank mRNA | AB209534.1 | - | 2 - 3 | 237 | 111 |
AJ890084.1 | - | 2 - 3 | 293 | 111 | |
AK025459.1 | - | 2 - 3 | 293 | 111 | |
AK298996.1 | - | 2 - 3 | 265 | 111 | |
AY040226.1 | - | 1 - 2 | 96 | 111 | |
BC009195.2 | - | 2 - 3 | 237 | 111 | |
BC066656.1 | - | 2 - 3 | 255 | 111 | |
EU794608.1 | - | 2 - 3 | 255 | 111 | |
FJ460515.1 | - | 2 - 3 | 255 | 111 | |
X15187.1 | - | 2 - 3 | 264 | 111 |