The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253379.1 | NP_001240308.1 | 3 - 4 | 707 | 89 |
NM_001253380.1 | NP_001240309.1 | 2 - 3 | 469 | 89 | |
NM_001253382.1 | NP_001240311.1 | 3 - 4 | 458 | 89 | |
NM_001253383.1 | NP_001240312.1 | 3 - 4 | 437 | 89 | |
NM_001253384.1 | NP_001240313.1 | 3 - 4 | 694 | 89 | |
NM_002948.3 | NP_002939.2 | 3 - 4 | 694 | 89 | |
GenBank mRNA | AF279903.1 | - | 2 - 3 | 348 | 89 |
AF283772.2 | - | 3 - 4 | 354 | 89 | |
AK091751.1 | - | 1 - 2 | 1434 | 89 | |
AK098795.1 | - | 3 - 4 | 344 | 89 | |
AK129815.1 | - | 3 - 4 | 354 | 89 | |
AK290694.1 | - | 3 - 4 | 350 | 89 | |
AK292000.1 | - | 3 - 4 | 350 | 89 | |
AK297091.1 | - | 3 - 4 | 352 | 89 | |
AY347528.1 | - | 2 - 3 | 419 | 89 | |
BC014837.1 | - | 3 - 4 | 357 | 89 | |
BC068198.1 | - | 1 - 1 | 353 | 89 | |
BC070328.1 | - | 3 - 4 | 350 | 89 | |
BC071672.1 | - | 1 - 1 | 339 | 89 | |
BC081565.1 | - | 3 - 4 | 359 | 89 | |
BE893727.1 | - | 3 - 4 | 429 | 89 | |
BT019672.1 | - | 2 - 3 | 314 | 89 | |
BU178524.1 | - | 3 - 4 | 458 | 89 | |
DA037869.1 | - | 3 - 4 | 375 | 89 | |
DA205054.1 | - | 3 - 4 | 420 | 89 | |
L25899.1 | - | 2 - 3 | 330 | 89 |