The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256062.2 | NP_001242991.1 | 1 - 2 | 1058 | 91 |
NM_001256321.2 | NP_001243250.1 | 2 - 3 | 659 | 91 | |
GenBank mRNA | BX378775.2 | - | 1 - 2 | 746 | 91 |
DB122722.1 | - | 2 - 3 | 370 | 91 |