The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270420.1 | NP_001257349.1 | 5 - 5 | 1226 | 98 |
NM_001270421.1 | NP_001257350.1 | 4 - 4 | 1116 | 98 | |
NM_032545.3 | NP_115934.1 | 6 - 6 | 1341 | 98 | |
GenBank mRNA | DQ786275.1 | - | 1 - 1 | 183 | 98 |