Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
-
Chromosome Location
Chr.1: 150272284 - 150280927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301039.1 NP_001287968.1 3 - 4 276 112
NM_001301040.1 NP_001287969.1 3 - 4 276 112
NM_001301042.1 NP_001287971.1 2 - 3 146 112
NM_024579.3 NP_078855.2 2 - 3 165 112
GenBank mRNA AA095731.1 - 2 - 3 133 112
AI342055.1 - 3 - 4 112
AK026874.1 - 2 - 3 150 112
AX770997.1 - 2 - 3 163 112
AX770998.1 - 3 - 4 190 112
AX770999.1 - 2 - 3 165 112
BC017761.1 - 2 - 3 150 112

View Full Product Details