Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM907830.1 - 3 - 4 405 Chr. 7 - 144252028 - 144256238 [-] on Build GRCh38 109


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101928605 LOC101928605 Anti-sense Chr. - 144195321 - 144299589 on Build GRCh38
Downstream olfactory receptor family 2 subfamily A member 7 OR2A7 HSDJ0798C17, OR2A21 4.0kb Sense Chr. - 144258696 - 144259628 on Build GRCh38

View Full Product Details