Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB480634.1 - 1 - 2 293 Chr. 8 - 62473217 - 62474269 [-] on Build GRCh38 115
GenBankmRNA DB480774.1 - 1 - 2 293 Chr. 8 - 62473288 - 62474269 [-] on Build GRCh38 115


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101929628 LOC101929628 -420.9kb Anti-sense Chr. - 61785072 - 62100736 on Build GRCh38
Downstream uncharacterized LOC643763 UG0898H09 503.6kb Anti-sense Chr. - 62977861 - 62984901 on Build GRCh38

View Full Product Details