The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_109956.1 | 6 - 7 | 525 | 81 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | Ral GTPase activating protein catalytic alpha subunit 2 | RALGAPA2 | AS250, C20orf74, bA287B20.1, dJ1049G11, dJ1049G11.4, p220 | -482.8kb | Sense | Chr. - 20389558 - 20712638 on Build GRCh38 |
Downstream | 5'-3' exoribonuclease 2 | XRN2 | 107.2kb | Anti-sense | Chr. - 21303284 - 21389825 on Build GRCh38 |