Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU154835.1 |
- |
1 - 2 |
189 |
|
Chr. 14 - 60515119 - 60554887 [+] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SIX homeobox 6 |
SIX6 |
MCOPCT2, ODRMD, OPTX2, Six9 |
-3.3kb
|
Sense |
Chr. - 60509220 - 60511807 on Build GRCh38 |
Downstream |
uncharacterized LOC105378189 |
LOC105378189 |
|
20.1kb
|
Anti-sense |
Chr. - 60571728 - 60643945 on Build GRCh38 |
View Full Product Details