Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB124028.1 |
- |
1 - 2 |
184 |
|
Chr. 2 - 241881948 - 241958936 [+] on Build GRCh38 |
98 |
GenBankmRNA |
AL702188.1 |
- |
1 - 2 |
99 |
|
Chr. 2 - 241882040 - 241934206 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1237 |
LINC01237 |
|
|
Sense |
Chr. - 241881363 - 242078722 on Build GRCh38 |
Downstream |
uncharacterized LOC285095 |
LOC285095 |
|
11.4kb
|
Anti-sense |
Chr. - 241893983 - 241902623 on Build GRCh38 |
View Full Product Details