Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4B
Entrez Gene ID
1975
Gene Name
eukaryotic translation initiation factor 4B
Gene Aliases
EIF-4B, PRO1843
Chromosome Location
Chr.12: 53006258 - 53042209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300821.1 NP_001287750.1 14 - 15 1999 76
NM_001417.5 NP_001408.2 14 - 15 1984 76

View Full Product Details