The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300898.1 | NP_001287827.1 | 4 - 5 | 738 | 112 |
NM_144643.3 | NP_653244.2 | 4 - 5 | 738 | 112 | |
XM_017007716.1 | XP_016863205.1 | 4 - 5 | 738 | 112 | |
XM_017007717.1 | XP_016863206.1 | 4 - 5 | 738 | 112 | |
XM_017007718.1 | XP_016863207.1 | 4 - 5 | 738 | 112 | |
XM_017007719.1 | XP_016863208.1 | 4 - 5 | 259 | 112 | |
XM_017007720.1 | XP_016863209.1 | 4 - 5 | 605 | 112 | |
GenBank mRNA | AK055217.1 | - | 4 - 5 | 738 | 112 |
BC121057.1 | - | 4 - 5 | 544 | 112 | |
BC121058.1 | - | 4 - 5 | 541 | 112 | |
BC128051.1 | - | 4 - 5 | 543 | 112 |