The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300933.1 | NP_001287862.1 | 2 - 3 | 450 | 90 |
GenBank mRNA | AY310328.1 | - | 2 - 3 | 404 | 90 |