The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300933.1 | NP_001287862.1 | 7 - 8 | 986 | 106 |
NM_001300934.1 | NP_001287863.1 | 5 - 6 | 821 | 106 | |
NM_001300935.1 | NP_001287864.1 | 7 - 8 | 1019 | 106 | |
NM_207172.1 | NP_997055.1 | 7 - 8 | 973 | 106 | |
NM_207173.1 | NP_997056.1 | 7 - 8 | 973 | 106 | |
GenBank mRNA | AY310326.1 | - | 7 - 8 | 973 | 106 |
AY310327.1 | - | 7 - 8 | 973 | 106 | |
AY310328.1 | - | 7 - 8 | 940 | 106 | |
AY310330.1 | - | 6 - 7 | 869 | 106 | |
AY310331.1 | - | 6 - 7 | 879 | 106 | |
AY310332.1 | - | 5 - 6 | 775 | 106 | |
AY956446.1 | - | 7 - 8 | 878 | 106 | |
AY956447.1 | - | 7 - 8 | 849 | 106 | |
AY956448.1 | - | 7 - 8 | 849 | 106 | |
BC132693.1 | - | 7 - 8 | 890 | 106 | |
BC132695.1 | - | 7 - 8 | 890 | 106 | |
BC144380.1 | - | 5 - 6 | 692 | 106 |