Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRP2
Entrez Gene ID
1466
Gene Name
cysteine and glycine rich protein 2
Gene Aliases
CRP2, LMO5, SmLIM
Chromosome Location
Chr.12: 76858715 - 76879040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300965.1 NP_001287894.1 5 - 6 585 116
NM_001321.2 NP_001312.1 5 - 6 610 116
GenBank mRNA AB209321.1 - 2 - 3 4762 116
BC000992.2 - 5 - 6 558 116
BT019913.1 - 4 - 5 506 116
U46006.1 - 4 - 5 506 116
U57646.1 - 5 - 6 562 116

View Full Product Details