The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003395.2 | NP_001003395.1 | 2 - 3 | 457 | 181 |
NM_001003396.2 | NP_001003396.1 | 2 - 3 | 355 | 181 | |
NM_001003397.2 | NP_001003397.1 | 2 - 3 | 355 | 181 | |
NM_001292026.2 | NP_001278955.1 | 2 - 3 | 457 | 181 | |
NM_001300994.2 | NP_001287923.1 | 2 - 3 | 355 | 181 | |
NM_001318903.1 | NP_001305832.1 | 2 - 3 | 355 | 181 | |
NM_001318907.1 | NP_001305836.1 | 2 - 3 | 252 | 181 | |
NM_003287.3 | NP_003278.1 | 2 - 3 | 355 | 181 | |
XM_005267122.2 | XP_005267179.1 | 2 - 3 | 136 | 181 | |
XM_006715557.3 | XP_006715620.1 | 2 - 3 | 601 | 181 | |
XM_017011237.1 | XP_016866726.1 | 2 - 3 | 601 | 181 | |
XM_017011238.1 | XP_016866727.1 | 2 - 3 | 601 | 181 | |
XM_017011239.1 | XP_016866728.1 | 2 - 3 | 457 | 181 | |
XM_017011240.1 | XP_016866729.1 | 2 - 3 | 135 | 181 | |
GenBank mRNA | AF004427.1 | - | 2 - 3 | 172 | 181 |
AF208012.1 | - | 2 - 3 | 171 | 181 | |
AK124817.1 | - | 2 - 3 | 457 | 181 | |
AK291866.1 | - | 2 - 3 | 319 | 181 | |
AK291872.1 | - | 2 - 3 | 189 | 181 | |
AK291887.1 | - | 2 - 3 | 150 | 181 | |
BC002375.2 | - | 2 - 3 | 245 | 181 | |
BE409865.1 | - | 2 - 3 | 383 | 181 | |
BG770463.1 | - | 2 - 3 | 253 | 181 | |
BM048527.1 | - | 2 - 3 | 375 | 182 | |
BM546629.1 | - | 2 - 3 | 228 | 181 | |
BP334548.1 | - | 2 - 3 | 271 | 181 | |
BQ641312.1 | - | 2 - 3 | 234 | 181 | |
CB114736.1 | - | 2 - 3 | 355 | 181 | |
U44427.1 | - | 2 - 3 | 312 | 181 |