The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301039.1 | NP_001287968.1 | 4 - 5 | 333 | 114 |
NM_001301040.1 | NP_001287969.1 | 4 - 5 | 333 | 114 | |
NM_001301041.1 | NP_001287970.1 | 2 - 3 | 119 | 114 | |
NM_001301042.1 | NP_001287971.1 | 3 - 4 | 203 | 114 | |
NM_024579.3 | NP_078855.2 | 3 - 4 | 222 | 114 | |
XM_011509984.2 | XP_011508286.1 | 3 - 4 | 249 | 114 | |
GenBank mRNA | AA095731.1 | - | 3 - 4 | 190 | 114 |
AI342055.1 | - | 2 - 3 | 114 | ||
AK026874.1 | - | 3 - 4 | 207 | 114 | |
AX770997.1 | - | 3 - 4 | 220 | 114 | |
AX770998.1 | - | 4 - 5 | 247 | 114 | |
AX770999.1 | - | 3 - 4 | 222 | 114 | |
BC017761.1 | - | 3 - 4 | 207 | 114 | |
DA358536.1 | - | 2 - 3 | 130 | 114 | |
W93038.1 | - | 3 - 4 | 114 |