Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN1
Entrez Gene ID
79906
Gene Name
MORN repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 2321253 - 2391751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301060.1 NP_001287989.1 8 - 9 965 112
NM_024848.2 NP_079124.1 8 - 9 965 112
XM_005244798.2 XP_005244855.1 7 - 8 829 112
XM_011542169.1 XP_011540471.1 8 - 9 965 112
XM_011542170.2 XP_011540472.1 8 - 9 965 112
XM_011542172.2 XP_011540474.1 7 - 8 829 112
XM_011542173.2 XP_011540475.1 8 - 9 965 112
XM_011542174.2 XP_011540476.1 8 - 9 965 112
XM_011542177.1 XP_011540479.1 8 - 9 965 112
XM_017002365.1 XP_016857854.1 8 - 9 965 112
GenBank mRNA AK024003.1 - 8 - 9 768 112
AK310323.1 - 7 - 8 829 112
BC021704.2 - 8 - 9 921 112

View Full Product Details