The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301167.1 | NP_001288096.1 | 4 - 5 | 491 | 69 |
NM_013252.2 | NP_037384.1 | 5 - 6 | 536 | 69 | |
XM_011515995.2 | XP_011514297.1 | 3 - 4 | 423 | 69 | |
XM_017011915.1 | XP_016867404.1 | 4 - 5 | 494 | 69 | |
XM_017011916.1 | XP_016867405.1 | 5 - 6 | 562 | 69 | |
XM_017011917.1 | XP_016867406.1 | 4 - 5 | 493 | 69 | |
GenBank mRNA | AJ271684.1 | - | 5 - 6 | 536 | 69 |
AK292766.1 | - | 5 - 6 | 525 | 69 | |
AK295200.1 | - | 4 - 5 | 428 | 69 | |
AK313703.1 | - | 5 - 6 | 525 | 69 | |
BC093714.1 | - | 4 - 5 | 360 | 69 | |
BC112099.1 | - | 4 - 5 | 360 | 69 | |
BC113098.1 | - | 5 - 6 | 478 | 69 | |
BC113099.1 | - | 4 - 5 | 409 | 69 | |
BC143392.1 | - | 3 - 4 | 291 | 69 |