The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185077.2 | NP_001172006.1 | 4 - 5 | 713 | 67 |
NM_001185078.2 | NP_001172007.1 | 4 - 5 | 483 | 67 | |
NM_001301240.1 | NP_001288169.1 | 4 - 5 | 483 | 67 | |
NM_001301241.1 | NP_001288170.1 | 4 - 5 | 713 | 67 | |
NM_001301242.1 | NP_001288171.1 | 4 - 5 | 483 | 67 | |
NM_001301243.1 | NP_001288172.1 | 3 - 4 | 618 | 67 | |
NM_004309.5 | NP_004300.1 | 4 - 5 | 483 | 67 | |
XM_011523574.1 | XP_011521876.1 | 3 - 4 | 843 | 67 | |
GenBank mRNA | AF498926.1 | - | 3 - 4 | 347 | 67 |
AK293147.1 | - | 4 - 5 | 427 | 67 | |
AK300816.1 | - | 4 - 5 | 456 | 67 | |
AK312347.1 | - | 4 - 5 | 419 | 67 | |
BC005851.2 | - | 4 - 5 | 439 | 67 | |
BC005875.2 | - | 4 - 5 | 439 | 67 | |
BC008701.3 | - | 3 - 4 | 374 | 67 | |
BC009759.2 | - | 4 - 5 | 404 | 67 | |
BC016031.1 | - | 3 - 4 | 355 | 67 | |
BC016185.1 | - | 4 - 5 | 399 | 67 | |
BC024258.1 | - | 4 - 5 | 399 | 67 | |
BC027730.2 | - | 4 - 5 | 402 | 67 | |
BC028333.1 | - | 4 - 5 | 402 | 67 | |
BC075827.1 | - | 4 - 5 | 399 | 67 | |
BC106044.1 | - | 4 - 5 | 442 | 67 | |
BI755343.1 | - | 4 - 5 | 404 | 67 | |
BP325393.1 | - | 4 - 5 | 414 | 67 | |
BQ674863.1 | - | 4 - 5 | 397 | 67 | |
BT006884.1 | - | 3 - 4 | 347 | 67 | |
CF132249.1 | - | 3 - 4 | 542 | 67 | |
CR456777.1 | - | 3 - 4 | 347 | 67 | |
D13989.1 | - | 3 - 4 | 367 | 67 | |
DC406556.1 | - | 4 - 5 | 483 | 67 | |
EU794615.1 | - | 3 - 4 | 374 | 67 | |
KU177916.1 | - | 3 - 4 | 347 | 67 | |
KU177917.1 | - | 3 - 4 | 347 | 67 | |
M97579.1 | - | 3 - 4 | 347 | 67 | |
X69550.1 | - | 4 - 5 | 400 | 67 |