The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005882.4 | NP_005873.2 | 3 - 4 | 517 | 97 |
GenBank mRNA | AF084928.1 | - | 4 - 5 | 465 | 97 |
AK022586.1 | - | 3 - 4 | 477 | 97 | |
AK308901.1 | - | 3 - 4 | 461 | 97 |