Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINJ2
Entrez Gene ID
4815
Gene Name
ninjurin 2
Gene Aliases
-
Chromosome Location
Chr.12: 564296 - 663741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016533.5 NP_057617.2 1 - 2 418 67
GenBank mRNA AF205633.1 - 1 - 2 93 67
AK296012.1 - 1 - 2 418 67
AK315995.1 - 1 - 2 416 67
BC057766.1 - 1 - 2 86 67
BI597990.1 - 1 - 2 271 67

View Full Product Details