Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR2
Entrez Gene ID
57047
Gene Name
phospholipid scramblase 2
Gene Aliases
-
Chromosome Location
Chr.3: 146391363 - 146497163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199978.1 NP_001186907.1 7 - 8 1139 94
NM_001199979.1 NP_001186908.1 5 - 6 756 94
NM_020359.2 NP_065092.1 6 - 7 869 94
XM_011513013.2 XP_011511315.2 5 - 6 1919 94
XM_011513019.2 XP_011511321.2 5 - 6 1919 94
XM_011513020.2 XP_011511322.2 5 - 6 1919 94
XM_011513021.2 XP_011511323.2 5 - 6 1919 94
XM_011513022.2 XP_011511324.2 5 - 6 1919 94
XM_011513023.2 XP_011511325.2 5 - 6 1919 94
XM_017006898.1 XP_016862387.1 5 - 6 1919 94
XM_017006899.1 XP_016862388.1 5 - 6 1919 94
XM_017006900.1 XP_016862389.1 5 - 6 1919 94
XM_017006901.1 XP_016862390.1 5 - 6 1919 94
XM_017006902.1 XP_016862391.1 5 - 6 1919 94
XM_017006903.1 XP_016862392.1 5 - 6 1919 94
XM_017006904.1 XP_016862393.1 5 - 6 1919 94
XM_017006905.1 XP_016862394.1 7 - 8 1375 94
XM_017006906.1 XP_016862395.1 7 - 8 1156 94
XM_017006907.1 XP_016862396.1 7 - 8 921 94
XM_017006908.1 XP_016862397.1 7 - 8 1046 94
XM_017006909.1 XP_016862398.1 6 - 7 833 94
XM_017006910.1 XP_016862399.1 5 - 6 946 94
XM_017006911.1 XP_016862400.1 6 - 7 1223 94
XM_017006912.1 XP_016862401.1 5 - 6 574 94
XM_017006914.1 XP_016862403.1 5 - 6 781 94
XM_017006915.1 XP_016862404.1 5 - 6 763 94
GenBank mRNA AF159441.1 - 6 - 7 813 94
AK301909.1 - 7 - 8 1139 94
BC055415.1 - 5 - 6 760 94
BC069785.1 - 5 - 6 706 94
BC120969.2 - 5 - 6 778 94
BC141969.1 - 5 - 6 778 94

View Full Product Details