Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM69
Entrez Gene ID
140691
Gene Name
tripartite motif containing 69
Gene Aliases
HSD-34, HSD34, RNF36, Trif
Chromosome Location
Chr.15: 44736362 - 44767829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301144.1 NP_001288073.1 3 - 4 687 98
NM_080745.4 NP_542783.2 3 - 4 679 98
NM_182985.4 NP_892030.3 4 - 5 1156 98
GenBank mRNA AF302088.1 - 3 - 4 535 98
AK226115.1 - 2 - 3 4943 98
AK292252.1 - 4 - 5 1000 98
AL360161.1 - 3 - 4 522 98
AY305385.1 - 4 - 5 854 98
BC024199.1 - 4 - 5 974 98
BC033314.1 - 3 - 4 475 98
BC047945.1 - 4 - 5 993 98

View Full Product Details