The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271706.1 | NP_001258635.1 | 1 - 2 | 252 | 151 |
XM_005251332.3 | XP_005251389.1 | 1 - 2 | 447 | 151 | |
XM_006716714.3 | XP_006716777.1 | 1 - 2 | 459 | 151 | |
GenBank mRNA | HY054657.1 | - | 1 - 2 | 252 | 151 |