The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272038.1 | NP_001258967.1 | 7 - 8 | 795 | 98 |
NM_002870.3 | NP_002861.1 | 7 - 8 | 677 | 98 | |
NR_073553.1 | 7 - 8 | 773 | 98 | ||
XM_017001959.1 | XP_016857448.1 | 6 - 7 | 430 | 98 | |
GenBank mRNA | AF498948.1 | - | 7 - 8 | 536 | 98 |
AK291580.1 | - | 7 - 8 | 596 | 98 | |
AY423722.1 | - | 7 - 8 | 537 | 98 | |
BC000799.1 | - | 7 - 8 | 630 | 98 | |
BC009227.2 | - | 1 - 1 | 513 | 98 | |
BC073168.1 | - | 7 - 8 | 581 | 98 | |
BC094846.1 | - | 7 - 8 | 661 | 98 | |
BT019700.1 | - | 7 - 8 | 536 | 98 | |
BT019701.1 | - | 7 - 8 | 536 | 98 | |
X75593.1 | - | 7 - 8 | 675 | 98 |