The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278063.1 | NP_001264992.1 | 1 - 2 | 2607 | 110 |
XM_006722525.3 | XP_006722588.1 | 1 - 2 | 2948 | 110 | |
GenBank mRNA | DW009659.1 | - | 1 - 2 | 110 |