The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278063.1 | NP_001264992.1 | 5 - 6 | 2915 | 95 |
XM_006722525.3 | XP_006722588.1 | 5 - 6 | 3256 | 95 |