Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI4
Entrez Gene ID
101928601
Gene Name
meiotic double-stranded break formation protein 4
Gene Aliases
-
Chromosome Location
Chr.6: 77650274 - 77927045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282136.2 NP_001269065.1 2 - 3 677 67
NM_001322247.1 NP_001309176.1 2 - 3 282 67
XM_017010155.1 XP_016865644.1 2 - 3 357 67

View Full Product Details