The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282136.2 | NP_001269065.1 | 4 - 5 | 1352 | 103 |
NM_001322247.1 | NP_001309176.1 | 4 - 5 | 957 | 103 | |
XM_005248774.3 | XP_005248831.1 | 3 - 4 | 730 | 103 | |
XM_017010155.1 | XP_016865644.1 | 4 - 5 | 1026 | 103 |