Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIXL1
Entrez Gene ID
83881
Gene Name
Mix paired-like homeobox
Gene Aliases
MILD1, MIX, MIXL
Chromosome Location
Chr.1: 226223618 - 226227055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282402.1 NP_001269331.1 1 - 2 458 107
GenBank mRNA BC143784.1 - 1 - 2 458 107

View Full Product Details