Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAQR4
Entrez Gene ID
124222
Gene Name
progestin and adipoQ receptor family member 4
Gene Aliases
-
Chromosome Location
Chr.16: 2969245 - 2973489 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284512.1 NP_001271441.1 1 - 2 599 72

View Full Product Details