The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286356.1 | NP_001273285.1 | 2 - 2 | 298 | |
NM_001286357.1 | NP_001273286.1 | 2 - 2 | 338 | 179 | |
NM_001286358.1 | NP_001273287.1 | 2 - 2 | 247 | ||
NM_002762.3 | NP_002753.2 | 2 - 2 | 254 | ||
GenBank mRNA | AK130957.1 | - | 2 - 2 | 254 | |
AK131573.1 | - | 2 - 2 | 298 | ||
AK307320.1 | - | 2 - 2 | 338 | 179 | |
BC042671.1 | - | 2 - 2 | 254 | ||
BC066338.1 | - | 2 - 2 | 254 | ||
DB066857.1 | - | 2 - 2 | 247 | ||
X07862.1 | - | 2 - 2 | 254 |