The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291316.1 | NP_001278245.1 | 3 - 4 | 424 | 96 |
GenBank mRNA | AB542951.1 | - | 3 - 4 | 370 | 96 |
BC144272.1 | - | 3 - 4 | 399 | 96 |