Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
Chromosome Location
Chr.5: 41925249 - 42040230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297437.1 NP_001284366.1 4 - 5 782 157
NM_012176.2 NP_036308.1 4 - 5 780 157
NM_033484.2 NP_277019.1 4 - 5 780 157
XM_006714470.3 XP_006714533.1 3 - 4 478 157
XM_011514026.2 XP_011512328.1 4 - 5 785 157
XM_011514027.2 XP_011512329.1 4 - 5 2155 157
GenBank mRNA AF176703.1 - 4 - 5 718 157
AK024075.1 - 4 - 5 780 157
BC040011.1 - 4 - 5 741 157
BC048098.1 - 4 - 5 748 157
CR749719.1 - 2 - 3 297 157

View Full Product Details