The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297565.1 | NP_001284494.1 | 3 - 4 | 387 | 101 |
GenBank mRNA | BC093060.1 | - | 3 - 4 | 302 | 101 |
BP366403.1 | - | 3 - 4 | 389 | 101 |