The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297753.1 | NP_001284682.1 | 4 - 5 | 902 | 74 |
XM_011526809.2 | XP_011525111.1 | 4 - 5 | 836 | 74 | |
XM_017026645.1 | XP_016882134.1 | 3 - 4 | 803 | 74 | |
GenBank mRNA | BC106047.1 | - | 4 - 5 | 902 | 74 |
BC143443.1 | - | 2 - 3 | 262 | 74 |