The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297773.1 | NP_001284702.1 | 5 - 6 | 1325 | 114 |
GenBank mRNA | M20879.1 | - | 5 - 6 | 1325 | 114 |