The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300832.1 | NP_001287761.1 | 7 - 8 | 951 | 124 |
NM_001300833.1 | NP_001287762.1 | 7 - 8 | 1017 | 124 | |
NM_031904.3 | NP_114110.1 | 8 - 9 | 1119 | 124 | |
GenBank mRNA | AF334946.1 | - | 9 - 10 | 1053 | 124 |
AK074850.1 | - | 7 - 8 | 884 | 124 | |
AK291678.1 | - | 8 - 9 | 1032 | 124 | |
AK304363.1 | - | 7 - 8 | 930 | 124 | |
BC044658.1 | - | 8 - 9 | 1037 | 124 | |
BC051695.1 | - | 8 - 9 | 1032 | 124 |