Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST6GALNAC6
Entrez Gene ID
30815
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases
SIAT7-F, SIAT7F, ST6GALNACVI
Chromosome Location
Chr.9: 127885321 - 127906725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104629.1 4 - 5 802 61
GenBank mRNA AK310850.1 - 4 - 5 802 61
KU178618.1 - 4 - 5 686 61


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL157554.1 - 2 - 3 422 Chr. 9 - 127886756 - 127894543 [-] on Build GRCh38 61
GenBankmRNA HY313062.1 - 2 - 3 Chr. 9 - 127886467 - 127890694 [+] on Build GRCh38 61
GenBankmRNA BQ723645.1 - 1 - 2 350 Chr. 9 - 127886395 - 127891005 [-] on Build GRCh38 61
GenBankmRNA BQ424678.1 - 2 - 3 546 Chr. 9 - 127886582 - 127894668 [-] on Build GRCh38 61
GenBankmRNA BQ719379.1 - 2 - 3 555 Chr. 9 - 127886611 - 127894677 [-] on Build GRCh38 61
GenBankmRNA DB179399.1 - 2 - 3 527 Chr. 9 - 127887561 - 127894649 [-] on Build GRCh38 61


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream adenylate kinase 1 AK1 HTL-S-58j -9.7kb Sense Chr. - 127866480 - 127886749 on Build GRCh38
Downstream ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 ST6GALNAC4 IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc 16.8kb Sense Chr. - 127907886 - 127917052 on Build GRCh38

View Full Product Details