Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GLIPR2
- Entrez Gene ID
-
152007
- Gene Name
- GLI pathogenesis related 2
- Gene Aliases
-
C9orf19, GAPR-1, GAPR1
- Chromosome Location
-
Chr.9: 36136536 - 36163913 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104640.1 |
|
2 - 3 |
387 |
117 |
GenBank mRNA |
DA824243.1 |
- |
2 - 3 |
156 |
117 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
reversion inducing cysteine rich protein with kazal motifs |
RECK |
ST15 |
-23.3kb
|
Sense |
Chr. - 36036905 - 36124455 on Build GRCh38 |
Downstream |
calicin |
CCIN |
BTBD20, KBTBD14 |
20.7kb
|
Sense |
Chr. - 36169392 - 36171334 on Build GRCh38 |
View Full Product Details