Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPD52
Entrez Gene ID
7163
Gene Name
tumor protein D52
Gene Aliases
D52, N8L, PC-1, PrLZ, hD52
Chromosome Location
Chr.8: 80034868 - 80171659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_105034.1 1 - 2 234 72
GenBank mRNA DB505575.1 - 1 - 2 132 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI667107.1 - 1 - 2 137 Chr. 8 - 80037878 - 80171556 [-] on Build GRCh38 72
GenBankmRNA HY180716.1 - 1 - 2 110 Chr. 8 - 80038183 - 80171535 [-] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream mitochondrial ribosomal protein S28 MRPS28 HSPC007, MRP-S28, MRP-S35, MRPS35 -23.0kb Sense Chr. - 79918860 - 80030271 on Build GRCh38
Downstream microRNA 5708 MIR5708 69.7kb Anti-sense Chr. - 80241389 - 80241473 on Build GRCh38

View Full Product Details