Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FBXL13
- Entrez Gene ID
-
222235
- Gene Name
- F-box and leucine rich repeat protein 13
- Gene Aliases
-
DRC6, Fbl13
- Chromosome Location
-
Chr.7: 102699226 - 103074841 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_105043.1 |
|
19 - 20 |
2113 |
88 |
GenBank mRNA |
BC026121.1 |
- |
19 - 20 |
2116 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RNA polymerase II subunit J2 |
POLR2J2 |
HRPB11B, RPB11b1 |
-150.3kb
|
Sense |
Chr. - 102636748 - 102671729 on Build GRCh38 |
Downstream |
leucine rich repeat containing 17 |
LRRC17 |
P37NB |
90.5kb
|
Anti-sense |
Chr. - 102912897 - 102945111 on Build GRCh38 |
View Full Product Details