Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01484
- Entrez Gene ID
-
101928136
- Gene Name
- long intergenic non-protein coding RNA 1484
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 173707599 - 173746209 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_108027.1 |
|
2 - 3 |
291 |
116 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
biorientation of chromosomes in cell division 1 |
BOD1 |
FAM44B |
-90.5kb
|
Sense |
Chr. - 173607145 - 173617077 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1485 |
LINC01485 |
|
74.8kb
|
Sense |
Chr. - 173786790 - 173790942 on Build GRCh38 |
View Full Product Details