Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CLIP4
- Entrez Gene ID
-
79745
- Gene Name
- CAP-Gly domain containing linker protein family member 4
- Gene Aliases
-
RSNL2
- Chromosome Location
-
Chr.2: 29097676 - 29183813 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109844.1 |
|
3 - 4 |
559 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 2 open reading frame 71 |
C2orf71 |
RP54 |
-56.4kb
|
Anti-sense |
Chr. - 29061690 - 29074261 on Build GRCh38 |
Downstream |
anaplastic lymphoma receptor tyrosine kinase |
ALK |
CD246, NBLST3 |
59.0kb
|
Anti-sense |
Chr. - 29192774 - 29921611 on Build GRCh38 |
View Full Product Details