Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC1A7
- Entrez Gene ID
-
6512
- Gene Name
- solute carrier family 1 member 7
- Gene Aliases
-
AAAT, EAAT5
- Chromosome Location
-
Chr.1: 53087179 - 53142632 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109858.1 |
|
7 - 8 |
1213 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
podocan |
PODN |
PCAN, SLRR5A |
-5.1kb
|
Anti-sense |
Chr. - 53062052 - 53085502 on Build GRCh38 |
Downstream |
carnitine palmitoyltransferase 2 |
CPT2 |
CPT1, CPTASE, IIAE4 |
103.6kb
|
Anti-sense |
Chr. - 53196429 - 53214197 on Build GRCh38 |
View Full Product Details