Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TYW5
- Entrez Gene ID
-
129450
- Gene Name
- tRNA-yW synthesizing protein 5
- Gene Aliases
-
C2orf60, hTYW5
- Chromosome Location
-
Chr.2: 199928911 - 199955736 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109905.1 |
|
3 - 4 |
566 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 2 open reading frame 69 |
C2orf69 |
|
-7.7kb
|
Anti-sense |
Chr. - 199911256 - 199928273 on Build GRCh38 |
Downstream |
chromosome 2 open reading frame 47 |
C2orf47 |
|
11.5kb
|
Anti-sense |
Chr. - 199955317 - 199964125 on Build GRCh38 |
View Full Product Details