Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CTC-436P18.1
- Entrez Gene ID
-
101928630
- Gene Name
- uncharacterized LOC101928630
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 61162316 - 61232080 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109908.1 |
|
3 - 4 |
422 |
110 |
GenBank mRNA |
BX109637.1 |
- |
3 - 4 |
422 |
110 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI218873.1 |
- |
1 - 2 |
|
|
Chr. 5 - 61179704 - 61232080 [-] on Build GRCh38 |
110 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small integral membrane protein 15 |
SMIM15 |
C5orf43 |
-17.9kb
|
Anti-sense |
Chr. - 61157709 - 61162475 on Build GRCh38 |
Downstream |
zinc finger SWIM-type containing 6 |
ZSWIM6 |
AFND |
100.2kb
|
Sense |
Chr. - 61332273 - 61546172 on Build GRCh38 |
View Full Product Details