The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_109975.1 | 3 - 4 | 437 | 146 | |
GenBank mRNA | BU568793.1 | - | 3 - 4 | 440 | 146 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | aryl hydrocarbon receptor nuclear translocator like 2 | ARNTL2 | BMAL2, CLIF, MOP9, PASD9, bHLHe6 | Anti-sense | Chr. - 27332854 - 27425813 on Build GRCh38 | |
Downstream | single-pass membrane protein with coiled-coil domains 2 | SMCO2 | C12orf70 | 56.3kb | Anti-sense | Chr. - 27446482 - 27506911 on Build GRCh38 |