Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101926955
Entrez Gene ID
101926955
Gene Name
uncharacterized LOC101926955
Gene Aliases
-
Chromosome Location
Chr.20: 25955812 - 25969288 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110003.1 2 - 3 179 75
GenBank mRNA BC052952.1 - 2 - 3 179 75


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI460299.1 - 318 Chr. - - [-] on Build 75
GenBankmRNA DB019897.1 - 2 - 3 157 Chr. 20 - 25955834 - 25965251 [+] on Build GRCh38 75
GenBankmRNA DB076627.1 - 1 - 2 355 Chr. 20 - 25955852 - 25965083 [+] on Build GRCh38 75
GenBankmRNA BU172390.1 - 3 - 4 345 Chr. 20 - 30228325 - 30259723 [+] on Build GRCh38 75
GenBankmRNA DB457853.1 - 310 Chr. - - [-] on Build 75
GenBankmRNA DB033375.1 - 1 - 2 359 Chr. 20 - 25955852 - 25965038 [+] on Build GRCh38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101926935 LOC101926935 -102.1kb Anti-sense Chr. - 25844667 - 25854021 on Build GRCh38
Downstream FSHD region gene 1 family member B, pseudogene FRG1BP C20orf80, FRG1B, bA348I14.2 117.5kb Sense Chr. - 30377203 - 30399331 on Build GRCh38

View Full Product Details